MAT184121884 Matrix Scientific

2-Piperidin-1-ylmethyl-morpholine

  • MDL number MFCD00770012
  • Cas Number : 122894-67-1
  • Molecular Weight : 184.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032626.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032626.html
Availability details for
Quantity
  • Availability
  • Add to Cart