MAT184121918 Matrix Scientific

2-Allyloxy-phenylamine

  • MDL number MFCD01902258
  • Cas Number : 27096-64-6
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032660.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032660.html
Availability details for
Quantity
  • Availability
  • Add to Cart