MAT184121920 Matrix Scientific

4-(2-Amino-2-methyl-propoxy)-furazan-3-ylaminehydrochloride

  • MDL number MFCD01221552
  • Cas Number : 1158514-88-5
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032662.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032662.html
Availability details for
Quantity
  • Availability
  • Add to Cart