MAT184121922 Matrix Scientific

3-(4-Hydroxy-8-methoxy-2-methyl-quinolin-3-yl)-propionic acid

  • MDL number MFCD01143209
  • Cas Number : 864433-55-6
  • Molecular Weight : 261.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032664.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032664.html
Availability details for
Quantity
  • Availability
  • Add to Cart