MAT184121943 Matrix Scientific

1-(2-Hydroxy-ethyl)-1,3-dihydro-benzoimidazol-2-one

  • MDL number MFCD02271217
  • Cas Number : 63388-01-2
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032685.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032685.html
Availability details for
Quantity
  • Availability
  • Add to Cart