MAT184121955 Matrix Scientific

Methyl-(1-phenyl-cyclopentylmethyl)-amine

  • MDL number MFCD06589807
  • Cas Number : 400756-68-5
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032697.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032697.html
Availability details for
Quantity
  • Availability
  • Add to Cart