MAT184121958 Matrix Scientific

4-(2-1H-Indol-3-yl-acetylamino)-butyric acid

  • MDL number MFCD01593582
  • Cas Number : 79438-89-4
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032700.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032700.html
Availability details for
Quantity
  • Availability
  • Add to Cart