MAT184121999 Matrix Scientific

4-(4-Methoxy-phenyl)-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine-6-carboxylic acid

  • MDL number MFCD00420637
  • Cas Number : 178324-01-1
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032741.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032741.html
Availability details for
Quantity
  • Availability
  • Add to Cart