MAT184122036 Matrix Scientific

[5-(2-Chloro-phenyl)-tetrazol-2-yl]-acetic acid

  • MDL number MFCD01029788
  • Cas Number : 312632-37-4
  • Molecular Weight : 238.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032778.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032778.html
Availability details for
Quantity
  • Availability
  • Add to Cart