MAT184122080 Matrix Scientific

2-o-Tolyl-benzooxazol-5-ylamine

  • MDL number MFCD00843562
  • Cas Number : 293737-82-3
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032822.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032822.html
Availability details for
Quantity
  • Availability
  • Add to Cart