MAT184122132 Matrix Scientific

(7-Amino-2,3-dihydro-benzo[1,4]dioxin-6-yl)-methanol

  • MDL number MFCD11053208
  • Cas Number : 1048922-42-4
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032874.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032874.html
Availability details for
Quantity
  • Availability
  • Add to Cart