MAT184122145 Matrix Scientific

2-Aminomethyl-5-tert-butyl-furan-3-carboxylicacid ethyl ester hydrochloride

  • MDL number MFCD02815368
  • Cas Number : 944467-98-5
  • Molecular Weight : 261.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125
Country of Origin USA
Product Link http://www.matrixscientific.com/032887.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032887.html
Availability details for
Quantity
  • Availability
  • Add to Cart