MAT184122176 Matrix Scientific

5-Ethyl-isoxazole-3-carboxylic acid

  • MDL number MFCD05864544
  • Cas Number : 52320-59-9
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032918.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032918.html
Availability details for
Quantity
  • Availability
  • Add to Cart