MAT184122178 Matrix Scientific

2-(5-Amino-2-hydroxymethyl-benzoimidazol-1-yl)-ethanol dihydrochloride

  • MDL number MFCD00434216
  • Cas Number : 1158316-59-6
  • Molecular Weight : 280.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032920.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032920.html
Availability details for
Quantity
  • Availability
  • Add to Cart