MAT184122193 Matrix Scientific

N-(3-Amino-phenyl)-2-morpholin-4-yl-acetamide

  • MDL number MFCD03044318
  • Cas Number : 462068-62-8
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032935.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032935.html
Availability details for
Quantity
  • Availability
  • Add to Cart