MAT184122205 Matrix Scientific

2-(2-Trifluoromethyl-benzoimidazol-1-yl)-butyric acid

  • MDL number MFCD05744840
  • Cas Number : 692764-48-0
  • Molecular Weight : 272.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032947.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032947.html
Availability details for
Quantity
  • Availability
  • Add to Cart