MAT184122222 Matrix Scientific

1-(4-Phenyl-cyclohexyl)-piperazine hydrochloride

  • MDL number MFCD11052937
  • Cas Number : 1052539-25-9
  • Molecular Weight : 280.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032964.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032964.html
Availability details for
Quantity
  • Availability
  • Add to Cart