MAT184122237 Matrix Scientific

2-[2-Ethyl-4-(2-methoxy-phenyl)-2-methyl-tetrahydro-pyran-4-yl]-ethylamine

  • MDL number MFCD01897900
  • Cas Number : 332392-22-0
  • Molecular Weight : 277.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032979.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032979.html
Availability details for
Quantity
  • Availability
  • Add to Cart