MAT184122273 Matrix Scientific

3-Methyl-7-nitro-1H-indole-2-carboxylic acid

  • MDL number MFCD03011739
  • Cas Number : 115058-18-9
  • Molecular Weight : 220.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033015.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033015.html
Availability details for
Quantity
  • Availability
  • Add to Cart