MAT184122312 Matrix Scientific

3-Fluoro-4-pyrrolidin-1-yl-benzaldehyde

  • MDL number MFCD05237677
  • Cas Number : 153659-43-9
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033054.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033054.html
Availability details for
Quantity
  • Availability
  • Add to Cart