MAT184122339 Matrix Scientific

2-Bromo-1-fluoro-4-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD11226531
  • Cas Number : 1099598-18-1
  • Molecular Weight : 257.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/032501.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032501.html
Availability details for
Quantity
  • Availability
  • Add to Cart