MAT184122344 Matrix Scientific

Furan-2-carboxylic acid (2-amino-benzothiazol-6-yl)-amide

  • MDL number MFCD03872201
  • Cas Number : 876709-45-4
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033086.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033086.html
Availability details for
Quantity
  • Availability
  • Add to Cart