MAT184122346 Matrix Scientific

4-(5-Amino-1-methyl-1H-benzoimidazol-2-yl)-butan-2-ol dihydrochloride

  • MDL number MFCD03872214
  • Cas Number : 1158490-09-5
  • Molecular Weight : 292.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033088.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033088.html
Availability details for
Quantity
  • Availability
  • Add to Cart