MAT184122363 Matrix Scientific

2-Chloro-N-(4-oxo-4,5-dihydro-thiazol-2-yl)-acetamide hydrochloride

  • MDL number MFCD11506452
  • Cas Number : 1185301-06-7
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033105.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033105.html
Availability details for
Quantity
  • Availability
  • Add to Cart