MAT184122391 Matrix Scientific

3-(5-Methyl-furan-2-yl)-phenylamine

  • MDL number MFCD01126401
  • Cas Number : 306935-67-1
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033133.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033133.html
Availability details for
Quantity
  • Availability
  • Add to Cart