MAT184122420 Matrix Scientific

2-(2-Chloro-phenoxy)-acetamidine hydrochloride

  • MDL number MFCD00173805
  • Cas Number : 58403-03-5
  • Molecular Weight : 221.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033162.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033162.html
Availability details for
Quantity
  • Availability
  • Add to Cart