MAT184122425 Matrix Scientific

3-Chloro-benzamidine hydrochloride

  • MDL number MFCD00276754
  • Cas Number : 24095-60-1
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033167.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033167.html
Availability details for
Quantity
  • Availability
  • Add to Cart