MAT184122437 Matrix Scientific

2-Morpholin-4-yl-2-pyridin-3-yl-ethylamine

  • MDL number MFCD01631945
  • Cas Number : 410544-52-4
  • Molecular Weight : 207.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033179.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033179.html
Availability details for
Quantity
  • Availability
  • Add to Cart