MAT184122448 Matrix Scientific

1-(2-Fluoro-benzyl)-1H-pyrazol-3-ylamine

  • MDL number MFCD02254004
  • Cas Number : 895929-38-1
  • Molecular Weight : 191.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033190.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033190.html
Availability details for
Quantity
  • Availability
  • Add to Cart