MAT184122452 Matrix Scientific

4-(4,5-Dimethoxy-2-methyl-phenyl)-thiazol-2-ylamine

  • MDL number MFCD02663981
  • Cas Number : 41213-87-0
  • Molecular Weight : 250.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033194.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033194.html
Availability details for
Quantity
  • Availability
  • Add to Cart