MAT184122465 Matrix Scientific

(7-Ethyl-2-methyl-1H-indol-3-yl)-acetic acid

  • MDL number MFCD02664375
  • Cas Number : 383131-65-5
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033207.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033207.html
Availability details for
Quantity
  • Availability
  • Add to Cart