MAT184122473 Matrix Scientific

4-Imidazol-1-yl-butylamine hydrochloride

  • MDL number MFCD11506474
  • Cas Number : 1185299-77-7
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033215.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033215.html
Availability details for
Quantity
  • Availability
  • Add to Cart