MAT184122496 Matrix Scientific

(3-Isobutyryl-indol-1-yl)-acetic acid

  • MDL number MFCD04392912
  • Cas Number : 797775-86-1
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033238.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033238.html
Availability details for
Quantity
  • Availability
  • Add to Cart