MAT184122547 Matrix Scientific

3-(4-Acetyl-3,5-dimethyl-pyrazol-1-yl)-propionic acid

  • MDL number MFCD06010628
  • Cas Number : 890596-67-5
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033289.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033289.html
Availability details for
Quantity
  • Availability
  • Add to Cart