MAT184122602 Matrix Scientific

2-Methyl-3-(2-methyl-imidazol-1-yl)-propionic acid

  • MDL number MFCD03834495
  • Cas Number : 696646-15-8
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033344.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033344.html
Availability details for
Quantity
  • Availability
  • Add to Cart