MAT184122625 Matrix Scientific

1-(4-Methoxy-phenoxy)-3-methylamino-propan-2-ol

  • MDL number MFCD05668681
  • Cas Number : 69336-80-7
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033367.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033367.html
Availability details for
Quantity
  • Availability
  • Add to Cart