MAT184122659 Matrix Scientific

2-(4-Fluoro-phenyl)-5-methyl-1H-imidazole-4-carboxylic acid

  • MDL number MFCD11053198
  • Cas Number : 933751-61-2
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033401.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033401.html
Availability details for
Quantity
  • Availability
  • Add to Cart