MAT184122665 Matrix Scientific

(4-Methyl-furazan-3-ylmethylsulfanyl)-acetic acid

  • MDL number MFCD07801110
  • Cas Number : 876715-34-3
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033407.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033407.html
Availability details for
Quantity
  • Availability
  • Add to Cart