MAT184122735 Matrix Scientific

2-(5,6-Dimethyl-1H-benzoimidazol-2-yl)-ethylaminehydrochloride

  • MDL number MFCD11506505
  • Cas Number : 1185294-80-7
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033427.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033427.html
Availability details for
Quantity
  • Availability
  • Add to Cart