MAT184122765 Matrix Scientific

4-Pyrazol-1-ylmethyl-phenylamine dihydrochloride

  • MDL number MFCD11506563
  • Cas Number : 1185134-73-9
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034025.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034025.html
Availability details for
Quantity
  • Availability
  • Add to Cart