MAT184122767 Matrix Scientific

1-(2-Piperidin-1-yl-ethyl)-[1,4]diazepanedihydrochloride

  • MDL number MFCD11506564
  • Cas Number : 1185293-20-2
  • Molecular Weight : 284.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034054.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034054.html
Availability details for
Quantity
  • Availability
  • Add to Cart