MAT184122788 Matrix Scientific

2-(2-Trifluoromethyl-benzoimidazol-1-yl)-propionic acid hydrochloride

  • MDL number MFCD11506596
  • Cas Number : 1219422-80-6
  • Molecular Weight : 294.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034315.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034315.html
Availability details for
Quantity
  • Availability
  • Add to Cart