MAT184122804 Matrix Scientific

3-Piperidin-1-yl-benzylamine

  • MDL number MFCD06659082
  • Cas Number : 175696-71-6
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/033484.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033484.html
Availability details for
Quantity
  • Availability
  • Add to Cart