MAT184122863 Matrix Scientific

4-Amino-1-phenethyl-pyrrolidin-2-one hydrochloride

  • MDL number MFCD11506523
  • Cas Number : 1177362-59-2
  • Molecular Weight : 240.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033543.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033543.html
Availability details for
Quantity
  • Availability
  • Add to Cart