MAT184122864 Matrix Scientific

4-Amino-1-(3-methoxy-phenyl)-pyrrolidin-2-onehydrochloride

  • MDL number MFCD11506524
  • Cas Number : 924855-21-0
  • Molecular Weight : 242.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033544.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033544.html
Availability details for
Quantity
  • Availability
  • Add to Cart