MAT184122875 Matrix Scientific

4-Aminomethyl-1-(3-methoxy-phenyl)-pyrrolidin-2-one

  • MDL number MFCD11500993
  • Cas Number : 1082279-12-6
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033426.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033426.html
Availability details for
Quantity
  • Availability
  • Add to Cart