MAT184122880 Matrix Scientific

3-p-Tolyl-pyrrolidin-3-ol hydrochloride

  • MDL number MFCD01687418
  • Cas Number : 67465-16-1
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033560.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033560.html
Availability details for
Quantity
  • Availability
  • Add to Cart