MAT184122881 Matrix Scientific

8-Ethoxy-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole

  • MDL number MFCD07364686
  • Cas Number : 876715-56-9
  • Molecular Weight : 216.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033415.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033415.html
Availability details for
Quantity
  • Availability
  • Add to Cart