MAT184122914 Matrix Scientific

2-Methyl-benzo[d]imidazo[2,1-b]thiazole-3-carbaldehyde

  • MDL number MFCD06824760
  • Cas Number : 876717-81-6
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033594.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033594.html
Availability details for
Quantity
  • Availability
  • Add to Cart