MAT184122925 Matrix Scientific

3-Amino-3-(3-chloro-4-methoxy-phenyl)-propionic acid

  • MDL number MFCD06206284
  • Cas Number : 773125-23-8
  • Molecular Weight : 229.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033605.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033605.html
Availability details for
Quantity
  • Availability
  • Add to Cart